siRNA Details
Gene Symbol: Fam150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109372.1 | NP_001102842.1 | 1, 2 | 699 | ||
GenBank mRNA | BC167018.1 | - | 1, 2 | 740 | 7307720 |

More Information
Other Species Targeted

