siRNA Details
Gene Symbol: Fam150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109372.1 | NP_001102842.1 | 3, 4 | 831 | ||
XM_006239971.2 | XP_006240033.1 | 2, 3 | 971 | |||
GenBank mRNA | BC167018.1 | - | 3, 4 | 872 | 7307720 |
