siRNA Details
Gene Symbol: Tex33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044289.1 | NP_001037754.1 | 2 | 612 | ||
GenBank mRNA | BC079191.1 | - | 2 | 612 | 7131327 |

More Information
Other Species Targeted

