siRNA Details
Gene Symbol: LOC685964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109489.1 | NP_001102959.1 | 2 | 181 | ||
XM_006236881.2 | XP_006236943.1 | 2 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
