siRNA Details
Gene Symbol: LOC685964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109489.1 | NP_001102959.1 | 2, 3 | 197 | ||
XM_006236881.2 | XP_006236943.1 | 2, 3 | 278 |

More Information
Other Species Targeted

