siRNA Details
Gene Symbol: LOC103691843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762006.1 | XP_008760228.1 | 53 |
Gene Symbol: LOC686900 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729188.2 | XP_002729234.2 | 1 | 53 |

More Information
Other Species Targeted

Gene Ontology Categories:
