siRNA Details
Gene Symbol: LOC102555635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246532.2 | XP_006246594.1 | 4 | 1695 | ||
XM_006246533.1 | XP_006246595.1 | 5 | 1578 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102555635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246532.2 | XP_006246594.1 | 4 | 1695 | ||
XM_006246533.1 | XP_006246595.1 | 5 | 1578 |