siRNA Details
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200045.1 | NP_001186974.1 | 10 | 1409 | ||
XM_008764610.1 | XP_008762832.1 | 9 | 1470 | |||
XM_008764611.1 | XP_008762833.1 | 10 | 3631 | |||
XM_008764612.1 | XP_008762834.1 | 8 | 1323 | |||
XM_008764613.1 | XP_008762835.1 | 8 | 1323 | |||
XM_008764614.1 | XP_008762836.1 | 8 | 1314 | |||
XM_008764615.1 | XP_008762837.1 | 8 | 1308 | |||
XM_008764616.1 | XP_008762838.1 | 9 | 1455 | |||
GenBank mRNA | AB558551.1 | - | 10 | 1409 | ||
AB558552.1 | - | 8 | 1030 | |||
FQ225774.1 | - | 6 | 690 |
