siRNA Details
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200045.1 | NP_001186974.1 | 9 | 1185 | ||
XM_008764610.1 | XP_008762832.1 | 8 | 1246 | |||
XM_008764611.1 | XP_008762833.1 | 9 | 3407 | |||
XM_008764612.1 | XP_008762834.1 | 7 | 1099 | |||
XM_008764613.1 | XP_008762835.1 | 7 | 1099 | |||
XM_008764614.1 | XP_008762836.1 | 7 | 1090 | |||
XM_008764615.1 | XP_008762837.1 | 7 | 1084 | |||
XM_008764616.1 | XP_008762838.1 | 8 | 1231 | |||
GenBank mRNA | AB558551.1 | - | 9 | 1185 | ||
AB558552.1 | - | 7 | 806 | |||
FQ225774.1 | - | 5 | 466 |
