siRNA Details
Gene Symbol: Atp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240845.2 | XP_006240907.1 | 1 | 47 | ||
GenBank mRNA | BC161836.1 | - | 1 | 57 | 9098303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
