siRNA Details
Gene Symbol: CUL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168370.1 | NP_001161842.1 | 22 | 4397 | ||
NM_014780.4 | NP_055595.2 | 22 | 4390 | |||
XM_005249503.2 | XP_005249560.1 | 22 | 4253 | |||
XM_006715285.1 | XP_006715348.1 | 22 | 4491 | |||
XM_011515019.2 | XP_011513321.1 | 22 | 4348 | |||
XM_011515020.2 | XP_011513322.1 | 22 | 4251 | |||
XM_011515021.1 | XP_011513323.1 | 17 | 2432 | |||
XM_017011533.1 | XP_016867022.1 | 22 | 4376 | |||
XM_017011534.1 | XP_016867023.1 | 22 | 4375 | |||
XM_017011535.1 | XP_016867024.1 | 22 | 4279 | |||
XM_017011536.1 | XP_016867025.1 | 22 | 4518 | |||
XM_017011537.1 | XP_016867026.1 | 22 | 4491 | |||
XM_017011538.1 | XP_016867027.1 | 22 | 4422 | |||
XM_017011539.1 | XP_016867028.1 | 22 | 4395 | |||
GenBank mRNA | AK292806.1 | - | 22 | 4327 | ||
AK302668.1 | - | 22 | 4334 | |||
AK307936.1 | - | 12 | 1818 | |||
BC033647.1 | - | 22 | 4372 | 5580027 | ||
D38548.1 | - | 22 | 4144 |
