siRNA Details
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109614.1 | NP_001103084.1 | 7 | 1019 | ||
XM_006241011.1 | XP_006241073.1 | 7 | 1066 | |||
GenBank mRNA | BC158787.1 | - | 7 | 977 | 7304442 | |
FQ219797.1 | - | 7 | 951 | |||
FQ220392.1 | - | 7 | 949 | |||
FQ229401.1 | - | 7 | 1066 | |||
FQ230290.1 | - | 7 | 954 | |||
FQ234550.1 | - | 7 | 999 |
