siRNA Details
Gene Symbol: Ccdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191063.1 | NP_001177992.1 | 11 | 1729 | ||
XM_008764384.1 | XP_008762606.1 | 12 | 1893 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191063.1 | NP_001177992.1 | 11 | 1729 | ||
XM_008764384.1 | XP_008762606.1 | 12 | 1893 |