siRNA Details
Gene Symbol: Lmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236972.2 | XP_006237034.1 | 3 | 1234 | ||
XM_006236973.2 | XP_006237035.1 | 3 | 1121 | |||
XM_575617.7 | XP_575617.4 | 2 | 1226 |

More Information
Other Species Targeted

Gene Ontology Categories:
