siRNA Details
Gene Symbol: Lrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109483.1 | NP_001102953.1 | 3 | 818 | ||
XM_008764719.1 | XP_008762941.1 | 2 | 836 |

More Information
Other Species Targeted

