siRNA Details
Gene Symbol: LOC691141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109624.1 | NP_001103094.1 | 7 | 701 | ||
XM_006242674.2 | XP_006242736.1 | 6 | 1939 | |||
XM_008765985.1 | XP_008764207.1 | 8 | 1999 | |||
XM_008765986.1 | XP_008764208.1 | 8 | 1999 | |||
GenBank mRNA | FQ227025.1 | - | 6 | 587 | ||
FQ230556.1 | - | 7 | 686 |
