siRNA Details
Gene Symbol: Vps37d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128193.2 | NP_001121665.1 | 4 | 1443 | ||
GenBank mRNA | BC166560.1 | - | 4 | 1443 | 7314601 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177574, NM_001199677, AY352519, BU704550, AY081953, and others.
