siRNA Details
Gene Symbol: LOC100910127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231301.1 | XP_006231363.1 | 5 | 796 |
Gene Symbol: LOC103691072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759203.1 | XP_008757425.1 | 5 | 796 |

More Information
Other Species Targeted

