siRNA Details
Gene Symbol: Mlkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772570.1 | XP_008770792.1 | 6, 7 | 1150 | ||
XM_008772571.1 | XP_008770793.1 | 6, 7 | 1105 | |||
XM_008772572.1 | XP_008770794.1 | 5, 6 | 1016 |
