siRNA Details
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109611.1 | NP_001103081.1 | 6 | 904 | ||
XM_006255634.2 | XP_006255696.1 | 6 | 1744 | |||
XM_006255635.2 | XP_006255697.1 | 6 | 1870 | |||
XM_006255636.2 | XP_006255698.1 | 6 | 1698 | |||
XM_006255638.1 | XP_006255700.1 | 5 | 749 | |||
XM_008772535.1 | XP_008770757.1 | 6 | 1744 |
