siRNA Details
Gene Symbol: LOC687295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748806.3 | XP_003748854.1 | 5 | 387 | ||
GenBank mRNA | FQ213435.1 | - | 4 | 148 | ||
FQ213942.1 | - | 4 | 148 | |||
FQ216794.1 | - | 4 | 148 |

More Information
Other Species Targeted

Gene Ontology Categories:
