siRNA Details
Gene Symbol: LOC687295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748806.3 | XP_003748854.1 | 9, 10 | 899 | ||
GenBank mRNA | FQ213435.1 | - | 8, 9 | 660 | ||
FQ213942.1 | - | 8, 9 | 660 | |||
FQ216794.1 | - | 8, 9 | 660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
