siRNA Details
Gene Symbol: RGD1561671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_341496.7 | XP_341497.4 | 1 | 189 |
Gene Symbol: Uqcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108528.1 | NP_001101998.1 | 2 | 230 | ||
XM_006256124.2 | XP_006256186.1 | 3 | 255 | |||
XM_008772733.1 | XP_008770955.1 | 3 | 679 | |||
GenBank mRNA | BC169127.1 | - | 397 | 6918056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
