siRNA Details
Gene Symbol: Brpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191572.1 | NP_001178501.1 | 8 | 2575 | ||
XM_006237055.2 | XP_006237117.1 | 8 | 2836 | |||
XM_006237056.2 | XP_006237118.1 | 8 | 2817 | |||
XM_006237057.2 | XP_006237119.1 | 8 | 2835 | |||
XM_006237058.2 | XP_006237120.1 | 8 | 2817 |

More Information
Other Species Targeted

Gene Ontology Categories:
