siRNA Details
Gene Symbol: Fam163b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109458.1 | NP_001102928.1 | 3 | 1081 | ||
XM_006233869.2 | XP_006233931.1 | 2 | 1014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
