siRNA Details
Gene Symbol: Brk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195476.1 | NP_001182405.1 | 2 | 166 | ||
GenBank mRNA | FM063685.1 | - | 2 | 154 | ||
FQ213048.1 | - | 2 | 166 | |||
FQ213578.1 | - | 2 | 166 | |||
FQ214114.1 | - | 2 | 166 | |||
FQ220806.1 | - | 2 | 164 | |||
FQ226823.1 | - | 2 | 166 | |||
FQ229010.1 | - | 2 | 166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
