siRNA Details
Gene Symbol: LOC687508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748835.3 | XP_003748883.1 | 4 | 244 | ||
XM_008759262.1 | XP_008757484.1 | 5 | 282 |
Gene Symbol: LOC100910772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748816.3 | XP_003748864.1 | 4 | 663 |
