siRNA Details
Gene Symbol: Pgm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191953.1 | NP_001178882.1 | 7 | 1125 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175013, AK035507.
Gene Ontology Categories:
