siRNA Details
Gene Symbol: LOC691921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135566.1 | NP_001129038.1 | 2, 3 | 165 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001135567, NM_001135569, NM_001135568, AK004515, BC038652, and others.
