siRNA Details
Gene Symbol: Mink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271136.1 | NP_001258065.1 | 12 | 1265 | ||
XM_006246716.2 | XP_006246778.1 | 12 | 1305 | |||
XM_006246717.2 | XP_006246779.1 | 12 | 1305 | |||
XM_006246718.2 | XP_006246780.1 | 12 | 1305 | |||
XM_006246720.2 | XP_006246782.1 | 12 | 1305 | |||
XM_006246721.2 | XP_006246783.1 | 12 | 1305 | |||
XM_006246722.2 | XP_006246784.1 | 12 | 1305 | |||
XM_006246723.2 | XP_006246785.1 | 12 | 1305 | |||
XM_006246724.2 | XP_006246786.1 | 12 | 1305 | |||
XM_006246725.2 | XP_006246787.1 | 11 | 1030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
