siRNA Details
Gene Symbol: Mink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271136.1 | NP_001258065.1 | 28 | 3497 | ||
XM_006246716.2 | XP_006246778.1 | 29 | 3627 | |||
XM_006246717.2 | XP_006246779.1 | 29 | 3624 | |||
XM_006246718.2 | XP_006246780.1 | 28 | 3603 | |||
XM_006246720.2 | XP_006246782.1 | 28 | 3540 | |||
XM_006246721.2 | XP_006246783.1 | 28 | 3516 | |||
XM_006246722.2 | XP_006246784.1 | 27 | 3516 | |||
XM_006246723.2 | XP_006246785.1 | 27 | 3429 | |||
XM_006246724.2 | XP_006246786.1 | 26 | 3405 | |||
XM_006246725.2 | XP_006246787.1 | 28 | 3352 | |||
GenBank mRNA | BC090005.1 | - | 3 | 315 | ||
BC103650.1 | - | 11 |

More Information
Other Species Targeted

Gene Ontology Categories:
