siRNA Details
Gene Symbol: Scimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001080739.4 | XP_001080739.2 | 3, 4 | 415 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001045526, AK138818, BC147178, BC147177.
Gene Ontology Categories:
