siRNA Details
Gene Symbol: Uqcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127553.2 | NP_001121025.1 | 3 | 190 | ||
GenBank mRNA | BC166882.1 | - | 3 | 175 | 6919310 | |
CB783276.1 | - | 3 | 190 | |||
FQ217398.1 | - | 3 | 175 | |||
FQ217669.1 | - | 3 | 181 | |||
FQ217799.1 | - | 3 | 189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
