siRNA Details
Gene Symbol: Pfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105791.1 | NP_001099261.1 | 4 | 373 | ||
XM_006246610.2 | XP_006246672.1 | 5 | 731 | |||
XM_006246611.2 | XP_006246673.1 | 4 | 554 | |||
XM_006246612.2 | XP_006246674.1 | 3 | 263 |

More Information
Other Species Targeted

Gene Ontology Categories:
