siRNA Details
Gene Symbol: Triap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126099.1 | NP_001119571.1 | 1 | 81 | ||
XM_006249468.2 | XP_006249530.1 | 1 | 92 | |||
GenBank mRNA | FQ230723.1 | - | 1 | 84 |
Gene Symbol: LOC100910137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751203.3 | XP_003751251.2 | 91 | |||
XM_006249488.2 | XP_006249550.1 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
