siRNA Details
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109645.2 | NP_001103115.1 | 2 | 164 | ||
XM_006234712.2 | XP_006234774.1 | 2 | 217 | |||
GenBank mRNA | BC161951.1 | - | 2 | 164 | 9026012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
