siRNA Details
Gene Symbol: LOC100910755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751957.3 | XP_003752005.1 | 2771 | |||
XM_008772948.1 | XP_008771170.1 | 2190 | ||||
XM_008772949.1 | XP_008771171.1 | 2188 |
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168586.1 | NP_001162058.1 | 17 | 1701 | ||
XM_006256392.2 | XP_006256454.1 | 17 | 2190 | |||
XM_006256393.2 | XP_006256455.1 | 17 | 2188 |

More Information
Other Species Targeted

Gene Ontology Categories:
