siRNA Details
Gene Symbol: Nipal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239277.2 | XP_006239339.1 | 12 | 1444 | ||
XM_578498.5 | XP_578498.3 | 12 | 1511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
