siRNA Details
Gene Symbol: Nipal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239277.2 | XP_006239339.1 | 12 | 1427 | ||
XM_578498.5 | XP_578498.3 | 12 | 1494 |

More Information
Other Species Targeted

Gene Ontology Categories:
