siRNA Details
Gene Symbol: Izumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229184.2 | XP_006229246.1 | 6 | 613 | ||
XM_008759451.1 | XP_008757673.1 | 5 | 592 | |||
XM_008759452.1 | XP_008757674.1 | 5 | 538 |
Gene Symbol: LOC100911976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228963.2 | XP_006229025.1 | 6 | 613 | ||
XM_008759429.1 | XP_008757651.1 | 5 | 592 | |||
XM_008759430.1 | XP_008757652.1 | 5 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
