siRNA Details
Gene Symbol: Lyc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128494.1 | NP_001121966.1 | 2 | 437 | ||
XM_006241392.2 | XP_006241454.1 | 2 | 261 | |||
GenBank mRNA | BC166737.1 | - | 2 | 437 | 7937244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
