siRNA Details
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234874.2 | XP_006234936.1 | 3 | 744 | ||
XM_006234875.2 | XP_006234937.1 | 3 | 744 | |||
XM_006234876.2 | XP_006234938.1 | 2 | 189 | |||
XM_006234877.1 | XP_006234939.1 | 2 | 182 | |||
XM_230512.7 | XP_230512.4 | 2 | 220 |
