siRNA Details
Gene Symbol: PJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014819.4 | NP_055634.3 | 4 | 714 | ||
XM_017010099.1 | XP_016865588.1 | 5 | 790 | |||
GenBank mRNA | AB007898.1 | - | 4 | 611 | ||
AK291759.1 | - | 4 | 648 | |||
BC030826.1 | - | 4 | 610 | 4811738 | ||
BQ228553.1 | - | 4 | 621 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
