siRNA Details
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109552.2 | NP_001103022.1 | 3 | 608 | ||
XM_008768114.1 | XP_008766336.1 | 2 | 703 | |||
GenBank mRNA | FM055064.1 | - | 3 | 561 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001078167, NM_173374, AK187083, AK169203, X66091, and others.
Gene Ontology Categories:
