siRNA Details
Gene Symbol: Ccdc182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277485.1 | NP_001264414.1 | 1 | 596 | ||
XM_006247075.2 | XP_006247137.1 | 1 | 1178 |

More Information
Other Species Targeted

Gene Ontology Categories:
