siRNA Details
Gene Symbol: Cox11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109575.1 | NP_001103045.1 | 639 |
Gene Symbol: LOC103693419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768168.1 | XP_008766390.1 | 3 | 684 |
