siRNA Details
Gene Symbol: Tcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271277.1 | NP_001258206.1 | 2 | 828 | ||
GenBank mRNA | FQ215695.1 | - | 2 | 826 | ||
FQ216780.1 | - | 2 | 828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
