siRNA Details
Gene Symbol: Pgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143895.1 | NP_001137367.1 | 6 | 693 | ||
XM_008768113.1 | XP_008766335.1 | 5 | 1117 | |||
GenBank mRNA | BC161914.1 | - | 6 | 660 | 8369613 |

More Information
Other Species Targeted

Gene Ontology Categories:
