siRNA Details
Gene Symbol: Tmem129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126274.1 | NP_001119746.1 | 1 | 138 | ||
GenBank mRNA | BC161857.1 | - | 1 | 138 | 9023916 |
Gene Symbol: LOC688176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751387.2 | XP_003751435.1 | 1 | 61 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026698, AK172044, BC094042, BC029088, AK002836.
Gene Ontology Categories:
