siRNA Details
Gene Symbol: Krtap3-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768204.1 | XP_008766426.1 | 1 | 280 |
Gene Symbol: Krtap3-3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247444.2 | XP_006247506.1 | 1 | 287 |
